Learn More
RUNX1T1 Mouse anti-Human, Unconjugated, Clone: 2G2F7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67086-1-IG-20UL
Description
The RUNX1T1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RUNX1T1. This antibody recognizes human, mouse, rat, pig antigen. The RUNX1T1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RUNX1T1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, ELISA | |
| 2G2F7 | |
| Unconjugated | |
| 20 μL | |
| 862 | |
| IgG1 |