missing translation for 'onlineSavingsMsg'
Learn More

RUNX1T1/ETO Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18379086 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
Unit Size:
25µL
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379086 25 μg 25µL
18356372 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18379086 Supplier Novus Biologicals Supplier No. NBP31868525UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RUNX1T1/ETO Polyclonal antibody specifically detects RUNX1T1/ETO in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX1T1/ETO
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100
Formulation PBS
Gene Accession No. Q06455
Gene Alias AML1T1protein CBFA2T1, CBFA2T1Cyclin-D-related protein, CDRMGC2796, core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclinD-related, DKFZp564B213, Eight twenty one protein, ETOFLJ33145, MTG8acute myelogenous leukemia 1 translocation 1, cyclin-D related, Protein ETO, Protein MTG8, runt-related transcription factor 1; translocated to, 1 (cyclin D-related), Zinc finger MYND domain-containing protein 2, ZMYND2myeloid translocation gene on 8q22
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against the RUNX1T1/ETO (RUNX1) fusion protein, using 3 different KLH-conjugated synthetic peptides. The antibody recognizes the ETO (RUNX1T1) part of the fusion protein.
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 862
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.