missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RUNX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA519638
This item is not returnable.
View return policy
Description
For Western Blot, this antibody has non-specific bands at 47 kDa and 75 kDa. This antibody is predicted to react with rat and chicken based on sequence homology.
RUNX1 belongs to the mammalian RUNX family of DNA binding proteins that regulate the expression of genes involved in cellular differentiation and cell cycle progression. RUNX1 is essential for hematopoietic stem cell generation in the vascular regions of the aorta, vitelline and umbilical arteries, yolk sac and placenta. RUNX1 is expressed in all tissues except brain and heart, with highest levels in thymus, bone marrow and peripheral blood.
Specifications
| RUNX1 | |
| Polyclonal | |
| Unconjugated | |
| RUNX1 | |
| acute myeloid leukemia 1 protein; Acute myeloid leukemia 2 protein; acute myeloid leukemia 3 protein; acute myeloid leukemia gene 2; AML1; AML-1; aml1 oncogene; AML1-EVI-1; AML1-EVI-1 fusion protein; AML2; AML3; AMLCR1; CBF2alpha; CBFA1; CBFA2; CBFA3; CBF-alpha 2; CBF-alpha-1; CBF-alpha-2; CBF-alpha-3; CCD; CCD1; CLCD; core binding factor alpha 2; core-binding factor runt domain alpha subunit 2 (acute myeloid leukemia 1 oncogene); Core-binding factor subunit alpha-1; Core-binding factor subunit alpha-2; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 1; core-binding factor, runt domain, alpha subunit 2; core-binding factor, runt domain, alpha subunit 3; EVI-1; Oncogene AML-1; oncogene AML-2; Oncogene AML-3; OSF2; OSF-2; osteoblast-specific transcription factor 2; OTTHUMP00000108697; OTTHUMP00000108700; OTTHUMP00000108702; PEA2 alpha C; PEA2aA; PEA2-alpha A; PEA2-alpha B; PEA2-alpha C; PEBP2 alpha C; PEBP2A; Pebp2a2; PEBP2A3; PEBP2aA; PEBP2aB; PEBP2aC; PEBP2alpha; PEBP2-alpha A; PEBP2-alpha B; PEBP2-alpha C; Pebpa2b; polyomavirus enhancer-binding protein 2 alpha A subunit; Polyomavirus enhancer-binding protein 2 alpha B subunit; polyomavirus enhancer-binding protein 2 alpha C subunit; RUN1; runt domain, alpha subunit 2; runt related transcription factor 1; runt related transcription factor 2; runt related transcription factor 3; runt-related transcription factor 1; Runt-related transcription factor 2; runt-related transcription factor 3; Runx1; RUNX1/ZNF687 fusion; RUNX2; RUNX3; SL3/AKV core-binding factor alpha A subunit; SL3/AKV core-binding factor alpha B subunit; SL3/AKV core-binding factor alpha C subunit; SL3-3 enhancer factor 1 alpha A subunit; SL3-3 enhancer factor 1 alpha B subunit; SL3-3 enhancer factor 1 alpha C subunit; transcription factor; transcription factor AML2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12394, 861 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.9 mg/mL | |
| PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| Q01196, Q03347 | |
| RUNX1 | |
| Synthetic peptide conjugated to KLH derived from within residues 400 to the C-terminus of Human RUNX1 / AML1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction