missing translation for 'onlineSavingsMsg'
Learn More

RUNX1/2/3 Rabbit anti-Human, Mouse, Clone: 6G5U2, Novus Biologicals™

Product Code. 18388302
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388302 100 μg 100µL
18371462 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18388302 Supplier Bio-Techne Supplier No. NBP316562100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RUNX1/2/3 Monoclonal antibody specifically detects RUNX1/2/3 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX1/2/3
Applications Western Blot
Classification Monoclonal
Clone 6G5U2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Acute myeloid leukemia 1, 2, 3 protein, AML1, 2, 3, AMLCR1, CBFA1, 2, 3, CBF-alpha-1, 2, 3, CCD, Core-binding factor subunit alpha-1, 2, 3, EVI-1, Oncogene AML-1, 2, 3, PEA2-alpha A, B, C, PEBP2-alpha A, B, C, Polyomavirus enhancer-binding protein 2 alpha A, B, C subunit, Runt-related transcription factor 1, 2, 3, RUNX family transcription factor 1, 2, 3, SL3/AKV core-binding factor alpha A, B, C subunit, SL3-3 enhancer factor 1 alpha A, B, C subunit
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 354-453 of human RUNX1/2/3 (Q01196).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Neuroscience, Protein Phosphatase, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 861
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.