missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3730 Running Buffer (10X)

Product Code. 15833550 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15833550 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15833550 Supplier Applied Biosystems™ Supplier No. 4335613
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The concentrated Running Buffer is used for electrophoresis on Applied Biosystems™3730 and 3730XL DNA Analyzers.

The concentrated Running Buffer is used for electrophoresis on Applied Biosystems™ 3730 and 3730xl DNA Analyzers.

Note: For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Running Buffer
Concentration 10 X
Content And Storage Store at 2°C to 8°C.
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer
For Use With (Application) Standard Sequencing
Product Type 3730 Running Buffer
Quantity 500 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.