missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RUNDC3B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563375

Product Code. 15731152

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: TPYLKYIQSS DSISSDEEEL RTLGSSGSES STPENVGPPF LMDENSWFNK CKRVKQKYQL TLEQKGYLEE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

RUNDC3B is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RUNDC3B
Polyclonal
Unconjugated
Rundc3b
Gm440; Rap2 binding protein 9; Rap2-binding protein 9; Rap2-interacting protein 9; RPIB9; RPIP9; RPIP-9; RUN domain containing 3B; RUN domain-containing protein 3B; rundc3b; zgc:101561
Rabbit
Antigen Affinity Chromatography
RUO
154661
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96NL0
Rundc3b
Recombinant protein corresponding to Human RUNDC3B. Recombinant protein control fragment (Product #RP-101732).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.