Learn More
Invitrogen™ RUNDC3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139934
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: NQLSESVSQN KILLQRIEDS DLAHKLEKEQ LEYIIVELQD QLTVLKNNDL RSRQELTAHL TNQWPSPGAL DVNAVALDTL LYRKHNKQW Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 100%.
RUNDC3B is a protein coding gene.
Specifications
| RUNDC3B | |
| Polyclonal | |
| Unconjugated | |
| Rundc3b | |
| Gm440; Rap2 binding protein 9; Rap2-binding protein 9; Rap2-interacting protein 9; RPIB9; RPIP9; RPIP-9; RUN domain containing 3B; RUN domain-containing protein 3B; rundc3b; zgc:101561 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 154661 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96NL0 | |
| Rundc3b | |
| Recombinant protein corresponding to Human RUNDC3B. Recombinant protein control fragment (Product #RP-109498). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.