missing translation for 'onlineSavingsMsg'
Learn More

Rumenic Acid (~80%), TRC

Product Code. 30137982
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137982 25 mg 25mg
30136461 50 mg 50mg
17908134 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137982 Supplier Toronto Research Chemicals Supplier No. TRCR70159025MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2540-56-9
Molecular Formula C18 H32 O2
Molecular Weight (g/mol) 280.45
Synonym 9,11-Octadecadienoic acid, (E,Z)- (8CI), (9Z,11E)-9,11-Octadecadienoic acid, (9Z,11E)-Octadecadienoic acid, 9-cis,11-trans-Linoleic acid, 9-cis,11-trans-Octadecadienoic acid, 9Z,11E-Octadecadienoic acid, Bovinic acid, Rumenic acid, cis-9,trans-11 Conjugated linoleic acid, cis-9,trans-11-Octadecadienoic acid, trans-11-cis-9-Octadecadienoic acid, 9,11-Octadecadienoic acid, (9Z,11E)-, Rumenic Acid
IUPAC Name (9Z,11E)-octadeca-9,11-dienoic acid
SMILES CCCCCC\C=C\C=C/CCCCCCCC(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h7-10H,2-6,11-17H2,1H3,(H,19,20)/b8-7+,10-9-
Formula Weight 280.2402
Chemical Name or Material Rumenic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.