missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUFY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19180-1-AP-20UL
This item is not returnable.
View return policy
Description
RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.Specifications
| RUFY2 | |
| Polyclonal | |
| Unconjugated | |
| RUFY2 | |
| KIAA1537, RABIP4R, RUFY2, ZFYVE13 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55680, 690777, 70432 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R4C2, Q8WXA3 | |
| RUFY2 | |
| RUFY2 Fusion Protein Ag5938 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction