missing translation for 'onlineSavingsMsg'
Learn More

Rufloxacin hydrochloride, MedChemExpress

Product Code. p-200098212
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16458496

Brand: Medchem Express HYB0902A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rufloxacin hydrochloride (MF-934 hydrochloride) is a fluoroquinolone antibacterial, inhibits B-cell differentiation in human mononuclear cells, inhibits Topo.

Rufloxacin hydrochloride (MF-934 hydrochloride) is a fluoroquinolone antibacterial, inhibits B-cell differentiation in human mononuclear cells, inhibits Topo.

Specifications

Chemical Name or Material Rufloxacin hydrochloride
Physical Form Solid
CAS 106017-08-7
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H19ClFN3O3S
For Use With (Application) COVID-19-immunoregulation
Synonym MF-934 hydrochloride
Solubility Information H2O : 2 mg/mL (5.00 mM; Need ultrasonic) ∣DMSO : 0.0852 mg/mL (0.21 mM; Need ultrasonic and warming)
SMILES O=C(C(C1=O)=CN2CCSC3=C(N4CCN(C)CC4)C(F)=CC1=C23)O.[H]Cl
Molecular Weight (g/mol) 399.87
Formula Weight 399.87
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado