missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rufloxacin hydrochloride, MedChemExpress
Description
Rufloxacin hydrochloride (MF-934 hydrochloride) is a fluoroquinolone antibacterial, inhibits B-cell differentiation in human mononuclear cells, inhibits Topo.
Specifications
Specifications
| Chemical Name or Material | Rufloxacin hydrochloride |
| Physical Form | Solid |
| CAS | 106017-08-7 |
| Color | Light Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H19ClFN3O3S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | MF-934 hydrochloride |
| Solubility Information | H2O : 2 mg/mL (5.00 mM; Need ultrasonic) ∣DMSO : 0.0852 mg/mL (0.21 mM; Need ultrasonic and warming) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?