missing translation for 'onlineSavingsMsg'
Learn More

Rufinamide, TRC

Product Code. 30006526
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006526 250 mg 250mg
30003910 50 mg 50mg
17903225 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006526 Supplier Toronto Research Chemicals Supplier No. TRCR701550250MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106308-44-5
Molecular Formula C10 H8 F2 N4 O
Molecular Weight (g/mol) 238.19
Synonym 1-[(2,6-Difluorophenyl)methyl]-1H-1,2,3-triazole-4-carboxamide, 1-[(2,6-Difiuorophenyl)methyl]-1H-1,2,3-triazole-4 carboxamide, Banzel, CGP 33101, E 2080, Inovelon, RUF 331, Rufinamide
IUPAC Name 1-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
SMILES NC(=O)c1cn(Cc2c(F)cccc2F)nn1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H8F2N4O/c11-7-2-1-3-8(12)6(7)4-16-5-9(10(13)17)14-15-16/h1-3,5H,4H2,(H2,13,17)
Formula Weight 238.0666
Percent Purity >95%
Chemical Name or Material Rufinamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.