missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Rufinamide
GREENER_CHOICE

Product Code. 18085082
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18085082 10 mg 10mg
18016002 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085082 Supplier Tocris Bioscience™ Supplier No. 3828/10
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Prolongs inactivation of sodium channels; anticonvulsant

Board spectrum anticonvulsant. Prolongs the inactivation of sodium channels and limits the frequency of action potential firing in cultured and acutely isolated neurons. Displays anticonvulsive activity in a range of animal seizure models.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106308-44-5
Molecular Formula C10H8F2N4O

Specifications

Quantity 10 mg
Formula Weight 238.19
Purity >99%
Molecular Weight (g/mol) 238.198
Chemical Name or Material Rufinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.