missing translation for 'onlineSavingsMsg'
Learn More

Rufinamide, MedChemExpress

Product Code. p-200100754
Encompass_Preferred
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
This item is not returnable. View return policy

Product Code. 16448278

Brand: Medchem Express HYA0042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rufinamide(E 2080; CGP 33101; RUF 331) is a new antiepileptic agent that differs structurally from other antiepileptic drugs and is approved as adjunctive therapy for Lennox-Gastaut syndrome (LGS).

Rufinamide(E 2080

Specifications

Chemical Name or Material Rufinamide
CAS 106308-44-5
Color White
Purity Grade Notes Research
Molecular Formula C10H8F2N4O
Quantity 10 mg
Synonym CGP 33101 E 2080 RUF 331
Solubility Information DMSO : 50 mg/mL (209.92 mM; Need ultrasonic)
SMILES O=C(C1=CN(CC2=C(F)C=CC=C2F)N=N1)N
Molecular Weight (g/mol) 238.19
Formula Weight 238.19
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.