missing translation for 'onlineSavingsMsg'
Learn More

Rufinamide, MedChemExpress

Product Code. 16448278
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448278 10 mg 10mg
16438006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448278 Supplier Medchem Express Supplier No. HYA0042
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rufinamide(E 2080; CGP 33101; RUF 331) is a new antiepileptic agent that differs structurally from other antiepileptic drugs and is approved as adjunctive therapy for Lennox-Gastaut syndrome (LGS).

Rufinamide(E 2080

Specifications

Chemical Name or Material Rufinamide
CAS 106308-44-5
Color White
Purity Grade Notes Research
Molecular Formula C10H8F2N4O
Quantity 10 mg
Synonym CGP 33101 E 2080 RUF 331
Solubility Information DMSO : 50 mg/mL (209.92 mM; Need ultrasonic)
SMILES O=C(C1=CN(CC2=C(F)C=CC=C2F)N=N1)N
Molecular Weight (g/mol) 238.19
Formula Weight 238.19
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.