missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rucaparib monocamsylate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Rucaparib (AG014699) monocamsylate is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP1. Rucaparib monocamsylate is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib monocamsylate has the potential for castration-resistant prostate cancer (CRPC) research.
Specifications
Specifications
| Chemical Name or Material | Rucaparib monocamsylate |
| CAS | 1859053-21-6 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C29H34FN3O5S |
| Quantity | 100 mg |
| Synonym | AG014699 monocamsylatePF-01367338 monocamsylate |
| Solubility Information | DMSO : 83.33 mg/mL (149.97 mM; Need ultrasonic) |
| SMILES | FC1=CC2=C3C(CCNC2=O)=C(C4=CC=C(CNC)C=C4)NC3=C1.O=S(C[C@@]5(C6(C)C)C(C[C@@]6([H])CC5)=O)(O)=O |
| Molecular Weight (g/mol) | 555.66 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction