missing translation for 'onlineSavingsMsg'
Learn More

Rubusoside, MedChemExpress

Product Code. 16474867
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474867 50 mg 50mg
16469058 10 mg 10mg
16429126 100 mg 100mg
16421478 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474867 Supplier Medchem Express Supplier No. HYN0668
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rubusoside is a natural sweetener and a solubilizing agent with antiangiogenic and antiallergic properties.

Rubusoside is a natural sweetener and a solubilizing agent with antiangiogenic and antiallergic properties.

Specifications

Chemical Name or Material Rubusoside
CAS 64849-39-4
Color White
Purity Grade Notes Research
Molecular Formula C32H50O13
Quantity 50 mg
Solubility Information DMSO : 110 mg/mL (171.14 mM; Need ultrasonic)
SMILES C[C@]([C@]1([H])CC2)(CCC[C@@]1(C)C(O[C@@H]([C@@H]([C@@H](O)[C@@H]3O)O)O[C@@H]3CO)=O)[C@@]4([H])[C@]2(CC5=C)C[C@@]5(O[C@]([C@@H]([C@@H](O)[C@@H]6O)O)([H])O[C@@H]6CO)CC4
Molecular Weight (g/mol) 642.73
Formula Weight 642.73
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.