missing translation for 'onlineSavingsMsg'
Learn More

Ruboxistaurin hydrochloride, MedChemExpress

Product Code. 16422816
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16422816

Brand: Medchem Express HY10195B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ruboxistaurin (LY333531) hydrochloride is an orally active, selective PKC beta inhibitor (Ki=2 nM). Ruboxistaurin hydrochloride exhibits ATP dependent competitive inhibition of PKC beta I with an IC50 of 4.7 nM. Ruboxistaurin hydrochloride inhibits PKC beta II with an IC50 of 5.9 nM.

Ruboxistaurin (LY333531) hydrochloride is an orally active, selective PKC beta inhibitor (Ki=2 nM). Ruboxistaurin hydrochloride exhibits ATP dependent competitive inhibition of PKC beta I with an IC50 of 4.7 nM. Ruboxistaurin hydrochloride inhibits PKC beta II with an IC50 of 5.9 nM.

Specifications

Chemical Name or Material Ruboxistaurin hydrochloride
CAS 169939-93-9
Color Red
Purity Grade Notes Research
Molecular Formula C28H29ClN4O3
Quantity 25 mg
Synonym LY333531 hydrochloride
Solubility Information DMSO : 6.67 mg/mL (13.21 mM; Need ultrasonic)
SMILES O=C1NC(C(C2=CN(C3=CC=CC=C32)CCO[C@H](CN(C)C)CC4)=C1C5=CN4C6=CC=CC=C56)=O.[H]Cl
Molecular Weight (g/mol) 505.01
Formula Weight 505.01
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.