missing translation for 'onlineSavingsMsg'
Learn More

uvex Rubiflex S XG35B Chemical Protection Gloves

Product Code. p-9618004 Shop All Uvex Products
Click to view available options
Size:
7
10
Unit Size:
10 pairs
Each
This item is not returnable. View return policy

Product Code. 17290441

Brand: uvex 6055707

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight, stockinette NBR chemical protection gloves feature Xtra Grip technology for enhanced grip in wet or oily areas.

  • Lightweight, stockinette NBR chemical protection gloves with XG Grip coating
  • Xtra Grip technology provides a secure grip in wet and oily areas
  • Cotton interlock supporting material ensures a high level of user comfort
  • Multi-layered construction provides mechanical abrasion resistance
  • Resistant to grease, mineral oils, and a wide variety of chemicals
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Laboratory
Coating Material NBR
Color Blue, Black
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 7
EN388 Standard 3121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKNOPT
For Use With (Application) Automotive, Chemical Handling, Laboratory Work, Maintenance, and Metal Handling
Gender Unisex
Glove Cuff Style Gauntlet
Liner Material Cotton
Series Rubiflex S XG35B
Length (Metric) Cuff 35 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.