missing translation for 'onlineSavingsMsg'
Learn More

uvex Rubiflex S NB709S Chemical Protection Gloves

Product Code. 17210421 Shop All Uvex Products
Change view
Click to view available options
Applications:
Laboratory
Unit Size:
10 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Applications Unit Size
17210421 Laboratory 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17210421 Supplier uvex Supplier No. 6024711
  • ONLINE EXCLUSIVE PRICE
    £515.00
    £47.92 / 10 pairs
    Save £467.08
    90% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

NBR-coated chemical protection gloves with reinforced cotton interlock supporting material, custom designed for fixed installation on sandblasting machines.

  • Special NBR-coated gloves with cotton interlock supporting material
  • Reinforced, anatomically shaped cuff provides additional protection
  • High water vapor absorption due to the cotton liner
  • Resistant to grease, mineral oils, and various chemicals
  • Custom designed for fixed installation on sandblasting machines
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Coating Material NBR
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 11
EN388 Standard 2121X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKOPT
For Use With (Application) Sandblasting
Gender Unisex
Liner Material Cotton
Series Rubiflex S NB709S
Length (Metric) 80 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.