Learn More
uvex Rubiflex S NB35SF Safety Glove
Product Code. p-7228477 Shop All Uvex Products
Click to view available options
Size:
8
9
10
11
Unit Size:
10 pairs
Product Code. 17787609
Brand: uvex 6020908
Only null left
Add to basket
Add to basket
The comfortable NBR safety glove, seamlessly coated in a reinforced design, with chemical and abrasion resistance.
  • Good heat/cold insulation on the palm
  • Abrasion and cut resistance
  • Ergonomic fit
  • High flexibility
  • Excellent mechanical characteristics
  • Good resistance to many chemicals, acids, alkalis, mineral oils and solvents
  • High water vapor absorption due to the cotton lining
  • Fully coated
  • Sandwich lining on palm, cuff
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN: 388 (2 1 2 1 X)
Color Green
Latex Free Yes
Material Cotton, Fleece, NBR
Product Type Chemical-resistant Gloves
Size 8