missing translation for 'onlineSavingsMsg'
Learn More

Rubidium tetrafluoroborate, 98%

Product Code. 11455746
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11455746

Brand: Thermo Scientific Alfa Aesar B20817.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18909-68-7 | BF4Rb | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18909-68-7
Molecular Formula BF4Rb
Molecular Weight (g/mol) 172.27
MDL Number MFCD00083071
InChI Key MERZTPVDEIGCOQ-UHFFFAOYSA-N
Synonym rubidium tetrafluoroborate, rubidium 1+ tetrafluoroborate, rubidium fluoroborate, rubidium boron fluoride, borate 1-, rubidium, acmc-1bu19, rubidium 1+ ion tetrafluoroborate
PubChem CID 23696894
IUPAC Name rubidium(1+) tetrafluoroboranuide
SMILES [Rb+].F[B-](F)(F)F

Specifications

Quantity 25 g
Linear Formula RbBF4
Formula Weight 172.27
Percent Purity 98%
Chemical Name or Material Rubidium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.