missing translation for 'onlineSavingsMsg'
Learn More

Rubidium sulfate, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number 11390719
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7488-54-2
H2O4RbS
183.54
MFCD00011190
XNNUAZGCOYTNQE-UHFFFAOYSA-N
197088
[Rb].OS(O)(=O)=O
This item is not returnable. View return policy
7488-54-2
H2O4RbS
183.54
MFCD00011190
XNNUAZGCOYTNQE-UHFFFAOYSA-N
197088
[Rb].OS(O)(=O)=O

Rubidium sulfate is used in hematology stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1050°C
Physical Form Powder
Quantity 50 g
Synonym rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate
Solubility Information Soluble in water.
IUPAC Name sulfuric acid rubidium
Formula Weight 267
Percent Purity 99.8%
Odor Odorless
Refractive Index 1.513
Density 3.613 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'