missing translation for 'onlineSavingsMsg'
Learn More

Rubidium nitrate, Puratronic™, 99.975% (metals basis)

Product Code. 11396997
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396997 10 g 10g
11386997 1 g 1g
11307007 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396997 Supplier Thermo Scientific Alfa Aesar Supplier No. 010852.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13126-12-0 | NO3Rb | 147.47 g/mol

Rubidium nitrate is used in infrared radiation producing pyrotechnic compositions. It acts as a colorant and an oxidizer especially in decoys and illumination flares. Further, it is employed as a starting material in the preparation of other rubidium compounds and rubidium metal, which is utilized for the preparation of catalysts and in scintillation counters. In addition to this, it is used in fireworks to produce a red-violet color.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13126-12-0
Molecular Formula NO3Rb
Molecular Weight (g/mol) 147.47
MDL Number MFCD00011193
InChI Key RTHYXYOJKHGZJT-UHFFFAOYSA-N
Synonym rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate
PubChem CID 25731
SMILES [Rb+].[O-][N+]([O-])=O

Specifications

Melting Point 310°C
Physical Form Crystalline
Quantity 10 g
UN Number UN1477
Solubility Information Soluble in water and nitric acid. Slightly soluble in acetone.
IUPAC Name rubidium(1+);nitrate
Formula Weight 147.47
Percent Purity 99.975%
Grade Puratronic
Refractive Index 1.52
Sensitivity Hygroscopic
Density 3.11 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium nitrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.