missing translation for 'onlineSavingsMsg'
Learn More

Rubidium nitrate, 99% (metals basis)

Product Code. 11336868
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336868 100 g 100g
11346868 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336868 Supplier Thermo Scientific Alfa Aesar Supplier No. 013496.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a catalyst in organic reactions and as chelating agents | CAS: 13126-12-0 | NO3Rb | 147.47 g/mol

Rubidium nitrate is used in infrared radiation producing pyrotechnic compositions as a colorant and an oxidizer, e.g. in decoys and illumination flares. It is also used as a raw material for preparation of other rubidium compounds and rubidium metal, for manufacture of catalysts and in scintillation counters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13126-12-0
Molecular Formula NO3Rb
Molecular Weight (g/mol) 147.47
MDL Number MFCD00011193
InChI Key RTHYXYOJKHGZJT-UHFFFAOYSA-N
Synonym rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate
PubChem CID 25731
SMILES [Rb+].[O-][N+]([O-])=O

Specifications

Melting Point 310°C
Color White
Physical Form Crystalline
Quantity 100 g
UN Number UN1477
Solubility Information Soluble in water; Slightly soluble in acetone; Very soluble in HNO3
IUPAC Name rubidium(1+) nitrate
Formula Weight 147.47
Percent Purity 99%
Refractive Index 1.52
Sensitivity Hygroscopic
Density 3.11 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium nitrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.