missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium nitrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Rubidium nitrate used in infrared radiation producing pyrotechnic compositions as a colorant and an oxidizer. It is also used as a raw material for preparation of other rubidium compounds and rubidium metal, for manufacture of catalysts and in scintillation counters. It is rarely used in fireworks to produce a red-violet colour.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 310°C |
Color | White |
Physical Form | Crystalline |
Quantity | 10 g |
UN Number | UN1477 |
Solubility Information | Soluble in water. Slightly soluble in acetone. Very soluble in nitric acid. |
Formula Weight | 147.47 |
Percent Purity | 99.8% |
Refractive Index | 1.52 |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'