missing translation for 'onlineSavingsMsg'
Learn More

Rubidium hydroxide hydrate, 99% (metals basis)

Product Code. 11326858
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11326858 5 g 5g
11306858 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11326858 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer 013481.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Oxidative chlorination catalyst | CAS: 12026-05-0 | HORb | 102.48 g/mol

Oxidative chlorination catalyst.Rubidium hydroxide hydrate is used in fireworks in place of pure rubidium to get violet color, thereby it prevents the waste of expensive element rubidium. It acts as an intermediate in the synthesis of other rubidium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 12026-05-0
Molecular Formula HORb
Molecular Weight (g/mol) 102.48
MDL Number MFCD00011194
InChI Key CPRMKOQKXYSDML-UHFFFAOYSA-M
Synonym rubidium hydroxide hydrate, rb.ho.h2o, rubidium hydroxide monohydrate, rubidium hydroxide-water 1/1/1, rubidium 1+ ion hydrate hydroxide, rubidium hydroxide rb oh , hydrate 9ci, rubidium hydroxide hydrate, fused solid-rb
PubChem CID 16212572
IUPAC Name rubidium(1+);hydroxide;hydrate
SMILES [OH-].[Rb+]

Specifikationer

Color White to Gray
Physical Form Crystalline
Quantity 5 g
Linear Formula RbOH·xH2O
UN Number UN2678
Merck Index 14,8285
Solubility Information Soluble in water and alcohol.
Formula Weight 102.48 (Anhydrous)
Percent Purity 99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rubidium hydroxide hydrate
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • Rubidium hydroxide hydrate
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faroangivelser
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Kompletterande information
  • MIXTURE LIST-Contain: Rubidium hydroxide hydrate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.