missing translation for 'onlineSavingsMsg'
Learn More

Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis)

Product Code. 11345799
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345799 50 g 50g
11335799 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345799 Supplier Thermo Scientific Alfa Aesar Supplier No. 010565.NE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-82-3 | HORb | 102.48 g/mol

Rubidium Hydroxide is used to give fireworks a violet color. It is a notable ntermediate in the synthesis of other rubidium compounds. It is used in photography and storage batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-82-3
Molecular Formula HORb
Molecular Weight (g/mol) 102.48
MDL Number MFCD00011194
InChI Key CPRMKOQKXYSDML-UHFFFAOYSA-M
Synonym rubidium hydroxide, rubidium hydroxide rb oh, rboh, rubidium 1+ hydroxide, rubidium hydroxide solution, rubidiumol, rubidium hydoxide, rubidium hydroxide sup 81 rb, rubidium monohydroxide, rb.ho
PubChem CID 62393
ChEBI CHEBI:32108
IUPAC Name rubidium(1+);hydroxide
SMILES [OH-].[Rb+]

Specifications

Color Yellow
Physical Form Liquid
Quantity 50 g
Linear Formula RbOH
UN Number UN2677
Merck Index 14,8285
Solubility Information Soluble in ethanol and water.
Formula Weight 102.48 (Anhydrous)
Percent Purity ≥99.6%
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Assay (metals basis)
Chemical Name or Material Rubidium hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.