missing translation for 'onlineSavingsMsg'
Learn More

Rubidium formate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11376868
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376868 10 g 10g
11386868 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376868 Supplier Thermo Scientific Alfa Aesar Supplier No. 013499.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rubidium formate hydrate is used in anticoagulants, dental materials, indicators and reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3495-35-0
Molecular Formula CHO2Rb
Molecular Weight (g/mol) 130.49
MDL Number MFCD00054359
InChI Key ZIMBPNXOLRMVGV-UHFFFAOYSA-M
Synonym rubidium formate, formic acid, rubidium salt, formic acid rubidium salt, 64-18-6 parent, formic acid, rubidium salt 1:1, rubidiumformiat, rubidium 1+ formira, rubidium 1+ formate, rubidium 1+ ion formate, formic acid, rubidiumsalt 1:1
PubChem CID 23673641
IUPAC Name rubidium(1+) formate
SMILES [Rb+].[O-]C=O

Specifications

Color White
Linear Formula RbOOCH·xH2O
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Miscible in water. Also miscible with ether,acetone,ethyl acetate,methanol and ethanol.
Formula Weight 130.49 (Anhydrous)
Percent Purity 99.8%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Rubidium formate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.