missing translation for 'onlineSavingsMsg'
Learn More

Rubidium fluoride, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals

Product Code. 11337607
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337607 2 g 2g
11327607 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337607 Supplier Thermo Scientific Alfa Aesar Supplier No. 012873.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of catalyst and for the manufacturing of high energy density micro cells and crystal scintillation counters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-74-7
Molecular Formula FRb
Molecular Weight (g/mol) 104.466
MDL Number MFCD00011191
InChI Key AHLATJUETSFVIM-UHFFFAOYSA-M
Synonym rubidium fluoride, rubidium fluoride rbf, rubidium monofluoride, rubidium 1+ fluoride, rubidium 1+ ion fluoride, rubidium fluoride, anhydrous, unii-63564tnj4r, acmc-20ajsd, rubidium i fluoride, rubidium fluoride, rbf
PubChem CID 83473
IUPAC Name rubidium(1+);fluoride
SMILES [F-].[Rb+]

Specifications

Melting Point 795°C
Color White
Physical Form Crystalline
Quantity 2 g
Linear Formula RbF
UN Number UN3288
Formula Weight 104.47
Percent Purity 99.975%
Grade Puratronic
Odor Odorless
Refractive Index 1.398
Sensitivity Hygroscopic
Density 3.557g/mL
Assay (metals basis)
Chemical Name or Material Rubidium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.