missing translation for 'onlineSavingsMsg'
Learn More

Rubidium fluoride, 99.1% (metals basis)

Product Code. 15402877
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 15402877

Brand: Thermo Scientific Alfa Aesar 047144.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-74-7 | FRb | 104.466 g/mol

It is used as raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of catalyst and for the manufacturing of high energy density micro cells and crystal scintillation counters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-74-7
Molecular Formula FRb
Molecular Weight (g/mol) 104.466
MDL Number MFCD00011191
InChI Key AHLATJUETSFVIM-UHFFFAOYSA-M
Synonym rubidium fluoride, rubidium fluoride rbf, rubidium monofluoride, rubidium 1+ fluoride, rubidium 1+ ion fluoride, rubidium fluoride, anhydrous, unii-63564tnj4r, acmc-20ajsd, rubidium i fluoride, rubidium fluoride, rbf
PubChem CID 83473
IUPAC Name rubidium(1+);fluoride
SMILES [F-].[Rb+]

Specifications

Melting Point 795°C
Color White
Physical Form Crystalline
Quantity 50 g
Linear Formula RbF
UN Number UN3288
Solubility Information Soluble in water. (1306g/L at 20°C).
Formula Weight 104.47
Percent Purity 99.1%
Refractive Index 1.398
Sensitivity Hygroscopic
Density 3.557 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium fluoride
Show More Show Less
compliance-icons
Product Identifier
  • Rubidium fluoride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Rubidium fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.