missing translation for 'onlineSavingsMsg'
Learn More

Rubidium chloride, Puratronic™, 99.975% (metals basis)

Product Code. 11376997
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376997 50 g 50g
11366997 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376997 Supplier Thermo Scientific Alfa Aesar Supplier No. 010851.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7791-11-9 | ClRb | 120.92 g/mol

Rubidium chloride is the mostly used rubidium compound, and finds its applications in various fields ranging from electrochemistry to molecular biology. It is used as a non-invasive biomarker, anergic and apathetic depressives in the field of biology; In addition, appropriate preparation of this salt is used as a radioactive isotope to study the perfusion of heart muscle. In the field of biochemistry, it is used to induce cells to take up DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+) chloride
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390°C
Melting Point 718°C
Color White
Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.975% (metals basis)
Solubility Information Soluble in water.
Formula Weight 120.92
Odor Odorless
Refractive Index 1.493
Sensitivity Hygroscopic
Appearance White crystals
Density 2.8
Chemical Name or Material Rubidium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.