missing translation for 'onlineSavingsMsg'
Learn More

Rubidium Chloride, Metal min. 99%., MP Biomedicals

Product Code. 11486202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
Unit Size:
100g
10g
50g
5g
Product Code. Quantity unitSize
11486202 5 g 5g
11496202 10 g 10g
11406212 50 g 50g
11416212 100 g 100g
4 options
This item is not returnable. View return policy

Product Code. 11486202

Brand: MP Biomedicals 0210282505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solutions may be slightly hazy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+) chloride
SMILES [Cl-].[Rb+]

Specifications

Quantity 5 g
Assay Percent Range Metal min. 99%.
Formula Weight 120.9
Chemical Name or Material Rubidium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.