missing translation for 'onlineSavingsMsg'
Learn More

Rubidium carbonate, Puratronic™, 99.975% (metals basis)

Product Code. p-4782101
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11326128

Brand: Thermo Scientific Alfa Aesar 011105.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 584-09-8 | CO3Rb2 | 230.94 g/mol

It finds it application as a raw material for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-09-8
Molecular Formula CO3Rb2
Molecular Weight (g/mol) 230.94
MDL Number MFCD00011188
InChI Key WPFGFHJALYCVMO-UHFFFAOYSA-L
Synonym rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate
PubChem CID 11431
SMILES [Rb+].[Rb+].[O-]C([O-])=O

Specifications

Melting Point 837°C
Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.975% (metals basis)
Solubility Information Soluble in water at 20°C 4500g/L.
IUPAC Name rubidium(1+);carbonate
Formula Weight 230.95
Odor Odorless
Sensitivity Very Hygroscopic
Appearance White powder
Chemical Name or Material Rubidium carbonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.