missing translation for 'onlineSavingsMsg'
Learn More

Rubidium bromide, 99.8% (metals basis)

Product Code. 11357627
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357627 25 g 25g
11347627 5 g 5g
11490310 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357627 Supplier Thermo Scientific Alfa Aesar Supplier No. 012891.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7789-39-1 | BrRb | 165.37 g/mol

Rubidium bromide is used in infrared optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-39-1
Molecular Formula BrRb
Molecular Weight (g/mol) 165.37
MDL Number MFCD00011186
InChI Key JAAGVIUFBAHDMA-UHFFFAOYSA-M
Synonym rubidium bromide, rubidium bromide rbbr, unii-33cm31xvqq, rubidium 1+ bromide, rubidium 1+ ion bromide, 33cm31xvqq, rubidium-bromid, rbbr, br.rb, rubidium i bromide
PubChem CID 4670918
IUPAC Name rubidium(1+);bromide
SMILES [Br-].[Rb+]

Specifications

Boiling Point 1340°C
Melting Point 693°C
Physical Form Crystalline
Quantity 25 g
Merck Index 14,8283
Solubility Information Soluble in water
Formula Weight 165.38
Percent Purity 99.8%
Odor Odorless
Refractive Index 1.553
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rubidium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.