missing translation for 'onlineSavingsMsg'
Learn More

Rubidium acetate, 99.8% (metals basis)

Product Code. 11327627
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 11327627

Brand: Thermo Scientific Alfa Aesar 012890.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 563-67-7 | C2H3O2Rb | 144.51 g/mol

Rubidium acetate has been reported to modify the acid/base character of the catalyst supports like silica, alumina, and titania. It can be used in the preparation of nanosized rubidium ferrite, and as a promoter on the palladium-catalyzed synthesis of vinyl acetate in heterogeneous gas reaction. It is used in medical and in research laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-67-7
Molecular Formula C2H3O2Rb
Molecular Weight (g/mol) 144.51
MDL Number MFCD00013058
InChI Key FOGKDYADEBOSPL-UHFFFAOYSA-M
Synonym rubidium acetate, acetic acid, rubidium salt, unii-86h795sz6d, acetic acid, rubidium salt 1:1, acetic acid rubidium, rubidio acetate, acmc-1aks3, rubidium 1+ acetate ion, rubidium 1+ ion acetate, c2h3o2.rb
PubChem CID 23673628
IUPAC Name rubidium(1+);acetate
SMILES [Rb+].CC([O-])=O

Specifications

Melting Point 246°C
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 144.52
Percent Purity 99.8%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Rubidium acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.