missing translation for 'onlineSavingsMsg'
Learn More

Rubidium, 99.75% (metals basis), Alfa Aesar™

Brand:  Alfa Aesar 010315.03

Code : NEW

Additional Details : CAS Number : 7440-17-7 Weight : 0.00100kg Transport : UN number : 1423 Chem class : 1 Pack group : I

Quantity 1g

 View more versions of this product

Product Code. 11371418

  • £82.30 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

Rubidium
Solid
MFCD00134055
7440-17-7
99.75% (Metals basis)

Specifications

Rubidium
7440-17-7
1.532g/cm3
99.75% (Metals basis)
Odorless
1g
Silver-colored solid
Solid
MFCD00134055
Silver
Videos
SDS