missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rubicon Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577807
This item is not returnable.
View return policy
Description
A 1:1,000 dilution of PA5-77807 was sufficient for detection of Rubicon in 15 μg of human Hela cell lysates by ECL immunoblot analysis using goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 95 kDa.
Two Beclin-1-interacting proteins, the run domain Beclin-1 interacting and cysteine-rich containing protein (Rubicon) and ATG14L, reciprocally regulate autophagy at different stages. Knockdown of Rubicon caused enhancement of autophagy while that of ATG14L caused a defect in autophagosome formation. Rubicon functions as part of a Beclin-1-PIK3C3-containing autophagy complex and is also an essential, positive regulator of the NAPDH oxidase complex. Upon microbial infection or TLR2 activation, Rubicon interacts with the CYBA subunit of the NAPDH oxidase complex, leading to a burst of reactive oxygen species and inflammatory cytokines.
Specifications
| Rubicon | |
| Polyclonal | |
| Unconjugated | |
| RUBCN | |
| 1700021K19Rik; 5330403K09; baron; Beclin-1 associated RUN domain containing protein; KIAA0226; mKIAA0226; RGD1305422; RIKEN cDNA 1700021K19 gene; RUBCN; Rubicon; RUN and cysteine rich domain containing beclin 1 interacting protein; RUN domain and cysteine-rich domain containing, Beclin 1-interacting protein; run domain Beclin-1 interacting and cysteine-rich containing protein; run domain Beclin-1 interacting and cystein-rich containing protein; run domain Beclin-1-interacting and cysteine-rich domain-containing protein; RUN domain protein as Beclin 1-interacting and cysteine-rich containing; rundataxin; SCAR15; similar to mKIAA0226 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9711 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| Q92622 | |
| RUBCN | |
| Synthetic peptide from the N-terminal of Human Rubicon. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction