missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rubicon Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562974
This item is not returnable.
View return policy
Description
Immunogen sequence: RSHSDTSIAS RGAPESCNDK AKLRGPLPYS GQSSEVSTPS SLYMEYEGGR YLCSGEGMFR RPSEGQSLIS YLSEQDFGSC ADLEKENAHF S Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 64%.
Two Beclin-1-interacting proteins, the run domain Beclin-1 interacting and cysteine-rich containing protein (Rubicon) and ATG14L, reciprocally regulate autophagy at different stages. Knockdown of Rubicon caused enhancement of autophagy while that of ATG14L caused a defect in autophagosome formation. Rubicon functions as part of a Beclin-1-PIK3C3-containing autophagy complex and is also an essential, positive regulator of the NAPDH oxidase complex. Upon microbial infection or TLR2 activation, Rubicon interacts with the CYBA subunit of the NAPDH oxidase complex, leading to a burst of reactive oxygen species and inflammatory cytokines.
Specifications
| Rubicon | |
| Polyclonal | |
| Unconjugated | |
| RUBCN | |
| 1700021K19Rik; 5330403K09; baron; Beclin-1 associated RUN domain containing protein; KIAA0226; mKIAA0226; RGD1305422; RIKEN cDNA 1700021K19 gene; RUBCN; Rubicon; RUN and cysteine rich domain containing beclin 1 interacting protein; RUN domain and cysteine-rich domain containing, Beclin 1-interacting protein; run domain Beclin-1 interacting and cysteine-rich containing protein; run domain Beclin-1 interacting and cystein-rich containing protein; run domain Beclin-1-interacting and cysteine-rich domain-containing protein; RUN domain protein as Beclin 1-interacting and cysteine-rich containing; rundataxin; SCAR15; similar to mKIAA0226 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9711 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92622 | |
| RUBCN | |
| Recombinant protein corresponding to Human Rubicon. Recombinant protein control fragment (Product #RP-100145). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction