missing translation for 'onlineSavingsMsg'
Learn More

Rubicon Antibody, DyLight 594, Novus Biologicals™

Product Code. 18429176 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429176 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429176 Supplier Novus Biologicals Supplier No. NBP281999DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rubicon Polyclonal antibody specifically detects Rubicon in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rubicon
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Baron, Beclin 1-interacting protein, Beclin-1 Associated RUN Domain Containing Protein, KIAA0226, Rubicon, RUN And Cysteine Rich Domain Containing Beclin 1 Interacting Protein, RUN Domain And Cysteine-Rich Domain Containing, Beclin 1-Interacting Protein, Run Domain Beclin-1-Interacting And Cysteine-Rich Domain-Containing Protein, rundataxin, SCAR15
Host Species Rabbit
Immunogen Rubicon antibody was raised against a 16 amino acid peptide near the amino terminus of human Rubicon. The immunogen is located within amino acids 240 - 290 of Rubicon. Amino Acid Squence: SGPPRKPQESRGHVSPAE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9711
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.