missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubicon Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-30461CL1
This item is not returnable.
View return policy
Description
Rubicon Polyclonal antibody specifically detects Rubicon in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Rubicon | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| Baron, Beclin 1-interacting protein, Beclin-1 Associated RUN Domain Containing Protein, KIAA0226, Rubicon, RUN And Cysteine Rich Domain Containing Beclin 1 Interacting Protein, RUN Domain And Cysteine-Rich Domain Containing, Beclin 1-Interacting Protein, Run Domain Beclin-1-Interacting And Cysteine-Rich Domain-Containing Protein, rundataxin, SCAR15 | |
| The immunogen for this product maps to a region between residue 922 and 972 of human Rubicon using the numbering given in entry NP_055502.1 (GeneID 9711). | |
| 0.1 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction | |
| 9711 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction