missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rubella Virus Virions Polyclonal Antibody, HRP
GREENER_CHOICE

Product Code. 11548451
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11548451 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11548451 Supplier Invitrogen™ Supplier No. PA173077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

The PA1-73077 antibody reacts with Rubella virus virions. PA1-73077 has been successfully used in ELISA applications. The PA1-73077 antibody was raised against Rubella virus strain HPV77.

Rubella virus was first isolated in 1962 by Parkman and Weller. Rubella virus is classified as a togavirus, genus Rubivirus. It is most closely related to group A arboviruses, such as Eastern and Western Equine Encephalitis viruses. It is an enveloped RNA virus, with a single antigenic type that does not cross-react with other members of the togavirus group. Rubella virus is relatively unstable and is inactivated by lipid solvents, trypsin, formalin, ultraviolet light, low pH, heat, and amantadine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rubella Virus Virions
Applications ELISA
Classification Polyclonal
Concentration 1-2 mg/mL
Conjugate HRP
Formulation PBS with 10mg/mL BSA and 0.002% thimerosal; pH 7.2
Host Species Goat
Immunogen Strain HPV77.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.