missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rubella Virus Virions Polyclonal Antibody, Biotin

Goat Polyclonal Antibody
Brand: Invitrogen™ PA173078
This item is not returnable.
View return policy
Description
The PA1-73078 antibody reacts with Rubella virus virions. PA1-73078 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73078 antibody was raised against Rubella virus strain HPV77.
Rubella virus was first isolated in 1962 by Parkman and Weller. Rubella virus is classified as a togavirus, genus Rubivirus. It is most closely related to group A arboviruses, such as Eastern and Western Equine Encephalitis viruses. It is an enveloped RNA virus, with a single antigenic type that does not cross-react with other members of the togavirus group. Rubella virus is relatively unstable and is inactivated by lipid solvents, trypsin, formalin, ultraviolet light, low pH, heat, and amantadine.
Specifications
| Rubella Virus Virions | |
| Polyclonal | |
| Biotin | |
| Goat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| 5.0 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Strain HPV77. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction