missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rubber Seal for Anaerobox Lid

Product Code. 15813197
Change view
Click to view available options
Includes:
Rectangular Seal
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15813197 Rectangular Seal 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15813197 Supplier Thermo Scientific™ Supplier No. AB0025S
  • ONLINE EXCLUSIVE PRICE
    £11.40
    £10.80 / 2.5 L
    Save £0.60
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seal only for 2.5L anaerobox lid AB0025L

Rubber seal for anaerobox lid. Seal only for 2.5L anaerobox lid AB0025L

Rubber seal for anaerobox lid. Seal only for 2.5L anaerobox lid AB0025L
TRUSTED_SUSTAINABILITY

Specifications

Description Seal only for 2.5L Anaerobox Lid
Includes Rectangular Seal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.