missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Rubber Pad

Product Code. 10583992 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Tubes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10583992 Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10583992 Supplier Thermo Scientific Supplier No. 76001805
  • ONLINE EXCLUSIVE PRICE
    £5.30
    £4.72 / Pack of 1
    Save £0.58
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rubber Pad for 50 mL and 100 mL tubes with round base.

  • Rubber Pad for 50 mL/100 mL tubes with round base
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Tubes
Disposable Reusable
Capacity 50 mL, 100 mL
Description Rubber Pad for 50 mL, 100 mL tubes with round base
Product Type Rubber Pad
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.