missing translation for 'onlineSavingsMsg'
Learn More

Progressive Safety™ Rubber Overshoes

Product Code. 12903035
Click to view available options
Size:
Medium
Large
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 12903035

Brand: Progressive Safety™ 2100EM

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Black
For Use With (Application) Convert ordinary shoes into anti-slip SRC rated footwear
Size Medium
Sole Features Grip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.