Learn More
ROBU Rubber Gaskets, GUKO
Product Code. 16674725
Click to view available options
Diameter (Metric)Top:
22 mm
40 mm
77 mm
Unit Size:
Pack of 5
Product Code. 16674725
Brand: ROBU 21851
Only null left
Add to basket
Add to basket
Conical gasket for filter funnels.
  • Conical
  • For filter funnels
Specifications
For Use With (Equipment) Filter Funnel
Material Natural Rubber
Quantity 5/Pk.
Diameter (Metric) Bottom 12 mm
Diameter (Metric)Top 22 mm