Learn More
Saint Gobain Life Sciences Rubber Gas Sample Bag
Use these bags for gas sample analysis. Saint Gobain Life Sciences Rubber Gas Sample Bags feature a cylindrical shape and an ebonite tube and tap.
Brand:  Saint Gobain Life Sciences 5607077
 View more versions of this product
Product Code. 11764498
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Bags are individually packaged
  • Available in packs of ten
  • Foil thickness: 5/10mm
  • Custom sizes and configurations available on request
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gas Sampling Bag
10 L
Faucet
Rubber