missing translation for 'onlineSavingsMsg'
Learn More

RU-SKI 43 hydrochloride, MedChemExpress

Product Code. 16409997
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16409997

Brand: Medchem Express HY18366A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RU-SKI 43 hydrochloride is a potent and selective Hedgehog acyltransferase (Hhat) inhibitor with an IC50 of 850 nM. RU-SKI 43 hydrochloride reduces Gli-1 activation through Smoothened-independent non-canonical signaling and decreases Akt and mTOR pathway activity. RU-SKI 43 hydrochloride has anti-cancer activity.

RU-SKI 43 hydrochloride is a potent and selective Hedgehog acyltransferase (Hhat) inhibitor with an IC50 of 850 nM. RU-SKI 43 hydrochloride reduces Gli-1 activation through Smoothened-independent non-canonical signaling and decreases Akt and mTOR pathway activity. RU-SKI 43 hydrochloride has anti-cancer activity.

Specifications

Chemical Name or Material RU-SKI 43 hydrochloride
CAS 1782573-67-4
Color White
Purity Grade Notes Research
Molecular Formula C22H31ClN2O2S
Quantity 5 mg
Solubility Information DMSO : ≥ 51 mg/mL (120.56 mM) ∣H2O : 2.5 mg/mL (5.91 mM; Need ultrasonic)
SMILES [H]Cl.O=C(CNCC(C)CC)N1CCC2=C(C=CS2)C1COC3=CC=CC(C)=C3
Molecular Weight (g/mol) 423.01
Formula Weight 423.01
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.