missing translation for 'onlineSavingsMsg'
Learn More

RU-SKI 43 hydrochloride, MedChemExpress

Product Code. 16443537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443537 50 mg 50mg
16417518 10 mg 10mg
16409997 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16443537 Supplier Medchem Express Supplier No. HY18366A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RU-SKI 43 hydrochloride is a potent and selective Hedgehog acyltransferase (Hhat) inhibitor with an IC50 of 850 nM. RU-SKI 43 hydrochloride reduces Gli-1 activation through Smoothened-independent non-canonical signaling and decreases Akt and mTOR pathway activity. RU-SKI 43 hydrochloride has anti-cancer activity.

RU-SKI 43 hydrochloride is a potent and selective Hedgehog acyltransferase (Hhat) inhibitor with an IC50 of 850 nM. RU-SKI 43 hydrochloride reduces Gli-1 activation through Smoothened-independent non-canonical signaling and decreases Akt and mTOR pathway activity. RU-SKI 43 hydrochloride has anti-cancer activity.

Specifications

Chemical Name or Material RU-SKI 43 hydrochloride
CAS 1782573-67-4
Color White
Purity Grade Notes Research
Molecular Formula C22H31ClN2O2S
Quantity 50 mg
Solubility Information DMSO : ≥ 51 mg/mL (120.56 mM) ∣H2O : 2.5 mg/mL (5.91 mM; Need ultrasonic)
SMILES [H]Cl.O=C(CNCC(C)CC)N1CCC2=C(C=CS2)C1COC3=CC=CC(C)=C3
Molecular Weight (g/mol) 423.01
Formula Weight 423.01
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.